Learn More
membrane frizzled-related protein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MFRP.
Marca: Abnova H00083552-A01.50uL
Dettagli aggiuntivi : Weight : 0.00010kg
Descrizione
This gene encodes a member of the frizzled-related proteins. The encoded protein may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen. The protein is encoded by a bicistronic mRNA, which also encodes C1q and tumor necrosis factor related protein 5
Sequence: NTTAFPNIWVGMITQEEVVEVLSGYKSLTSLPCYQHFRRLLCGLLVPRCTPLGSVLPPCRSVCQEAEHQCQSGLALLGTPWPFNCNRLPEAADLEACAQPSpecifica
membrane frizzled-related protein | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MFRP. | |
MFRP | |
C1QTNF5/FLJ30570/NNO2/rd6 | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_031433 | |
MFRP | |
MFRP (NP_113621, 480 a.a. ∼ 579 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
83552 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |