missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCOR Monoclonal Antibody (731229), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524345
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
LCOR Monoclonal antibody specifically detects LCOR in Human samples. It is validated for Immunocytochemistry.Specifications
LCOR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96JN0 | |
LCOR | |
E. coli-derived recombinant human LCoR Asn110-Gln338 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry | |
731229 | |
Unconjugated | |
LCOR | |
3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2 | |
Mouse | |
Protein A/G | |
RUO | |
84458 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |