missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
KIT, Human, Matched Antibody Pair (AP43), Abnova™
To detect and quantify protein level of human
Brand: Abnova H00003815-AP43.1Set
Indeholder: Capture antibody: mouse monoclonal anti-KIT (100μg) , Detection antibody: biotinylated mouse monoclonal anti-KIT, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Tekniske data
| KIT | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KIT. | |
| C-Kit/CD117/PBT/SCFR | |
| Mouse | |
| RUO | |
| 3815 | |
| Human | |
| Liquid |
| ELISA | |
| Biotin | |
| KIT | |
| KIT | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-KIT (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-KIT, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion