Få mere at vide
anti-KHK, Human, Matched Antibody Pair, Abnova™
Artikelnummer. 16102284
Skift visning
Mængde:
1 Set
Pakningsstørrelse:
1 sæt
Artikelnummer. Mængde unitSize
16102284 1 Set 1 sæt
1 options
Artikelnummer. 16102284 Leverandør Abnova Leverandørnr. H00003795AP41.1Set
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody: mouse monoclonal anti-KHK, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-KHK, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Tekniske data
Antigen KHK
Applikationer ELISA
Klassifikation Monoclonal/Polyclonal
Konjugeret Biotin
Oversigt This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KHK.
Gene KHK
Gensymboler KHK
Værtsarter Mouse
Mængde 1 Set
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 3795
Inkluderer Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-KHK, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-KHK, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Målarter Human
Indhold og opbevaring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Vis mere Vis mindre
For Research Use Only