missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ IRAK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517531
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
It is not recommended to aliquot this antibody.
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
Tekniske data
| IRAK4 | |
| Polyclonal | |
| Unconjugated | |
| IRAK4 | |
| 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 266632, 300177, 51135 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 300 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q8R4K2, Q9NWZ3 | |
| IRAK4 | |
| Synthetic peptide corresponding to residues surrounding lysine 41 of human IRAK4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion