missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Millipore™ anti-Integrin αVβ5, Clone: P1F6
Brand: Merck™ MAB1961
Additional Details : Weight : 0.10000kg
Description
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Specifications
Integrin αVβ5 | |
Semi-purified | |
Human UCLA P3 cells (lung carcinoma) | |
100μg | |
Cell Structure | |
Monoclonal |
Unconjugated | |
Murine | |
IgG1 | |
RUO | |
Primary | |
Chicken, Human |