missing translation for 'onlineSavingsMsg'
Learn More
Learn More
inner membrane protein, mitochondrial (mitofilin), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IMMT.
Brand: Abnova H00010989-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: TQLRRQAAAHTDHLRDVLRVQEQELKSEFEQNLSEKLSEQELQFRRLSQEQVDNFTLDINTAYARLRGIEQAVQSHAVAEEEARKAHQLWLSVEALKYSMSpecifications
inner membrane protein, mitochondrial (mitofilin) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant IMMT. | |
IMMT | |
DKFZp779P1653/HMP/MGC111146/P87/P87/89/P89/PIG4/PIG52 | |
Mouse | |
50 μL | |
Yes | |
10989 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006839 | |
IMMT | |
IMMT (NP_006830, 481 a.a. ∼ 580 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |