missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Rat anti-Mouse, APC-Cy7, Clone: JES6-5H4, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 560547

1248.69 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15860289

  • 1250.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2) and has been reported to be neutralizing.

Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse IL-2 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-2
Monoclonal
0.2mg/mL
Il2; Interleukin-2; T-cell growth factor; TCGF
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
JES6-5H4
APC-Cyanine7
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse IL-2 Recombinant Protein
50 μg
Immunology
Mouse
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IL-2 Rat anti-Mouse, APC-Cy7, Clone: JES6-5H4, BD > 50μg; APC-Cy7

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.