missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ IL-13 Monoclonal Antibody (JES10-5A2)
Rat Monoclonal Antibody
Brand: Invitrogen™ AHC0132
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-13 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA, Neu
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Tekniske data
| IL-13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35225 | |
| Il13 | |
| Purified recombinant human IL-13. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot | |
| JES10-5A2 | |
| Unconjugated | |
| Il13 | |
| ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
| Rat | |
| Purified | |
| RUO | |
| 3596 | |
| -20°C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion