missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK beta Monoclonal Antibody (10AG2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516154
Additional Details : Weight : 0.10000kg
Description
Suggested positive control: Daudi, HeLa, NIH-3T3.
IKK beta Monoclonal antibody specifically detects IKK beta in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
IKK beta | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.02% sodium azide | |
O14920, O88351, Q9QY78 | |
Ikbkb | |
full-length human IKKbeta recombinant protein | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
10AG2 | |
Unconjugated | |
Ikbkb | |
AI132552; AIM-1; FLJ40509; I-kappa-B kinase 2; I-kappa-B-kinase beta; Ikbkb; IKK[b]; IKK2; IKK-2; IKKB; IKK-B; IKKbeta; IKK-beta; IMD15; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; inhibitor of nuclear factor kappa-B kinase subunit beta; MGC131801; NFKBIKB; nuclear factor NF-kappa-B inhibitor kinase beta | |
Mouse | |
Protein G | |
RUO | |
16150, 3551, 84351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |