Learn More
anti-IgG2a κ, Rat, FITC, Clone: R35-95, Isotype Control, BD
1695.00 DKK - 2290.00 DKK
Especificaciones
Clone | R35-95 |
---|---|
Concentration | 0.5mg/mL |
Applications | Flow Cytometry, Western Blot |
Conjugate | FITC |
Host Species | Rat |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15887838
|
BD Biosciences
554688 |
0.1 mg |
1695.00 DKK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10187143
|
BD Biosciences
553929 |
0.25 mg |
2290.00 DKK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Descripción
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Especificaciones
R35-95 | |
Flow Cytometry, Western Blot | |
Rat | |
IgG2a κ | |
Undiluted at 4°C |
0.5mg/mL | |
FITC | |
Mouse Pooled Immunoglobulin | |
Affinity Purified |