Få mere at vide
anti-IgG2a κ, Mouse, Alexa Fluor™ 647, Clone: G155-178, BD
Brand: BD Biosciences 557715
Beskrivelse
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Tekniske data
| Flow Cytometry | |
| Alexa Fluor 647 | |
| TNP-keyhole limpet hemocyanin | |
| 100 prøver | |
| IgG2a κ |
| G155-178 | |
| Mus | |
| Affinity Purified | |
| Undiluted at 4°C |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.