missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1, kappa, BB515, Clone: X40, BD
Brand: BD Biosciences 564416
Additional Details : Weight : 0.01000kg
Description
Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Immunogen: Keyhole limpet hemocyanin (KLH)
Flow Cytometry, Isotype control
Specifications
IgG1 κ | |
Brilliant Blue 515 (BB515) | |
Keyhole limpet hemocyanin (KLH) | |
100 μg |
X40 | |
Mouse | |
Affinity Purified | |
IgG1 κ |