Få mere at vide
IFNE, Human, IP-WB Antibody Pair (PW2), Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: mouse monoclonal anti-IFNE (300μg), Antibody pair for WB: rabbit polyclonal anti-IFNE (50μL)
Tekniske data
Tekniske data
| Antigen | IFNE |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | IFNE |
| Gene Alias | IFN-E/IFNE1/IFNT1/MGC119018/MGC119020/PRO655 |
| Gensymboler | IFNE |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only