missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG (H+L) Secondary Antibody, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen 628400

Additional Details : Weight : 0.50000kg

Product Code. 10353143

  • 1340.00 DKK / Each
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Reconstitute with 1 mL of distilled water.

Human IgG (H+L) Polyclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Human IgG (H+L)
Polyclonal
Unconjugated
Goat
Affinity chromatography
RUO
Human
Lyophilized
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.4
Gamma Immunoglobin
1 mL
Secondary
4° C
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers