missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen A24478
Additional Details : Weight : 0.01000kg
Description
Human IgG Fc Cross-Adsorbed Polyclonal antibody specifically detects Human IgG Fc Cross-Adsorbed in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Human IgG Fc Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobin Fc region | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |