Learn More
Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A18799
Additional Details : Weight : 0.01000kg
Description
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin human serum immunoglobulins, light chains on all human immunoglobulins, serum proteins from bovine, mouse or rabbit.
Human IgE Cross-Adsorbed Polyclonal antibody specifically detects Human IgE Cross-Adsorbed in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Human IgE Cross-Adsorbed | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
Human IgE | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |