Få mere at vide
Hsp70 Mouse, Unlabeled, Clone: 7, BD
Beskrivelse
Hsp70 is a member of the heat shock or stress-induced genes whose expression is regulated at both transcriptional and translational levels. Elevated temperatures induce a transient resistance to subsequent heat exposure in various types of cells. Although Hsp70 protein is constitutively expressed at low levels, its expression abundantly increases in response to heat stress. During such stress, Hsp70 leaves the cytoplasm and nucleoplasm and rapidly accumulates in the nucleolus. Removal of the stress results in the redistribution of Hsp70 to its original locations.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Tekniske data
Tekniske data
| Antigen | Hsp70 |
| Applikationer | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 7 |
| Koncentration | 250μg/mL |
| Konjugeret | Unconjugated |
| Formulering | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Oprensningsmetode | Affinity Purified |
| Vis mere |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.