Få mere at vide
anti-HLA-E, Human, IP-WB Antibody Pair, Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit purified polyclonal anti-HLA-E (300μg), Antibody pair for WB: mouse purified polyclonal anti-HLA-E (50μg)
Tekniske data
Tekniske data
| Antigen | HLA-E |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | HLA-E |
| Gene Alias | DKFZp686P19218/EA1.2/EA2.1/HLA-6.2/MHC/QA1 |
| Gensymboler | HLA-E |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only