Få mere at vide
HLA-DR, DP, DQ Mouse anti-Human, FITC, Clone: TÜ39, BD
Beskrivelse
Reacts with a monomorphic epitope of major histocompatibility Class II HLA-DR antigens expressed on B cells, activated T cells, and antigen presenting cells. Detects the αβ complex and not the isolated chains. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Tekniske data
Tekniske data
| Antigen | HLA-DR, DP, DQ |
| Applikationer | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | TÜ39 |
| Konjugeret | FITC |
| Oversigt | Tü39 |
| Formulering | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Oprensningsmetode | Affinity Purified |
| Mængde | 100 Tests |
| Vis mere |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.