missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Tag Monoclonal Antibody (L5), Alexa Fluor™ 647, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA1142A647
Additional Details : Weight : 0.00900kg
Description
similar epitope as the Flag tag from Sigma.
DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Western Blot.Specifications
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Rat | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
L5 | |
Alexa Fluor 647 | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
DYKDDDDK-tagged mouse Langerin | |
50 μL | |
Primary | |
Antibody | |
IgG2a |