Få mere at vide
FIV p24, clone: vpg 50, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125653
Beskrivelse
The lentivirus FIV is responsible for a progressive and debilitating immunodeficiency syndrome in domestic cats, similar to that caused by HIV. It is a complex retrovirus with a tightly-packed genome containing the structrual genes gag, env and pro-pol, as well as accessory genes vif, rev and ORF-A/2.
MA1-25653 detects FIV p24 from FIV p24 samples.MA1-25653 has been successfully used in Western blot procedures.The MA1-25653 immunogen is recombinant p24.
Tekniske data
| FIV p24 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Supernatant |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| vpg 50 | |
| Feline Immunodeficiency Virus | |
| Recombinant p24. | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.