Learn More
FDPS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528228
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, H1299, HCT116, Mouse brain, rat liver. Predicted reactivity: Mouse (89%), Rat (89%), Pig (89%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FDPS Polyclonal antibody specifically detects FDPS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FDPS | |
Polyclonal | |
Unconjugated | |
FDPS | |
(2E,6E)-farnesyl diphosphate synthase; 6030492I17Rik; Ac2-125; AI256750; cholesterol-regulated 39 kDa protein; CR 39; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; Fdps; Fdpsl1; FPP synthase; FPP synthetase; FPPS; FPS; geranyltranstransferase; KIAA1293; mKIAA1293; POROK9; testis-specific farnesyl pyrophosphate synthetase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110196, 2224, 83791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.75 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P05369, P14324, Q920E5 | |
FDPS | |
Recombinant fragment corresponding to a region within amino acids 43 and 381 of Human FDPS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |