missing translation for 'onlineSavingsMsg'
Få mere at vide

anti-FADD, EZR Protein Protein Interaction Antibody Pair, Abnova™

Artikelnummer. 16012478
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 Set
Pakningsstørrelse:
1 sæt
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16012478 1 Set 1 sæt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16012478 Leverandør Abnova Leverandørnr. DI0170.1Set

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Indeholder

FADD rabbit purified polyclonal antibody (20μg); EZR mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FADD protein, and the other against the EZR protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Tekniske data

Antigen FADD
Applikationer In situ PLA
Klassifikation Monoclonal/Polyclonal
Konjugeret Unconjugated
Gene FADD
Gene Alias GIG3/MGC8528/MORT1
Gensymboler FADD
Værtsarter Mouse, Rabbit
Mængde 1 Set
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 8772/7430
Inkluderer Antibody pair set content: 1. FADD rabbit purified polyclonal antibody (20 ug) 2. EZR mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Målarter Human
Indhold og opbevaring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Produkttype FADD & EZR Protein Protein Interaction Antibody Pair
Form Purified
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.