missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Monoclonal Antibody (9035), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA143037
Additional Details : Weight : 0.01000kg
Description
MA1-43037 detects Factor VIII in mouse samples. MA1-43037 has successfully been used in Western blot and ELISA applications. MA1-43037 was derived from mouse factor VIII.
Factor VIII Monoclonal antibody specifically detects Factor VIII in Mouse samples. It is validated for ELISA, Western Blot.Specifications
Factor VIII | |
Monoclonal | |
8.1 mg/mL | |
50% water with 50% glycerol and no preservative | |
Q06194 | |
F8 | |
Mouse Factor VIII. | |
100 μg | |
Primary | |
Mouse | |
Antibody |
ELISA, Western Blot | |
9035 | |
Unconjugated | |
F8 | |
AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7 | |
Rat | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
14069 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |