missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Estrone Rabbit anti-Chemical, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA185741
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PA1-85741 is specific for oestrone. Less than 0.01% cross-reactivity was observed with oestradiol 17B, oestriol, progesterone and testosterone. Rabbit anti estrone antibody recognizes the aromatized C18 steroid, estrone.
Estrone is an aromatized C18 steroid with a 3-hydroxyl group and a 17-ketone, a major mammalian estrogen. it is converted from androstenedione directly, or from testosterone via estradiol. In humans, it is produced primarily by the cyclic ovaries, placenta, and the adipose tissue of men and postmenopausal women. Therapeutic estrone is the synthetic form of a naturally occurring estrogen estrone. Estrone diffuses through the cell membrane and binds to and subsequently activates the nuclear estrogen receptor found in the reproductive tract, breast, pituitary, hypothalamus, liver, and bone. The activated hormone-receptor complex gets translocated to the nucleus where it binds to the estrogen response element on the dna and promotes the transcription of target genes involved in the functioning of the female reproductive system and secondary sex characteristics. Estrone increases the hepatic synthesis of sex hormone binding globulin (shbg), thyroid-binding globulin (tbg), and other serum proteins, as well as suppresses the release of follicle-stimulating hormone (fsh) and luteinizing hormone (lh) from the anterior pituitary. [National Center for Biotechnology Information. PubChem Compound Database; CID=5870]
Tekniske data
| Estrone | |
| Polyclonal | |
| Folliculin; OESTRONE; C18H22O2 | |
| Synthetic oestrone conjugated to BSA. | |
| RUO | |
| 4°C or -20°C if preferred | |
| Liquid | |
| Chemical |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion