Learn More
erythrocyte membrane protein band 4.2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human EPB42 protein.
Brand: Abnova H00002038-B01.50uL
Additional Details : Weight : 0.00010kg
Description
Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MGQALGIKSCDFQAARNNEEHHTKALSSRRLFVRRGQPFTIILYFRAPVRAFLPALKKVALTAQTGEQPSKINRTQATFPISSLGDRKWWSAVVEERDAQSWTISVTTPADAVIGHYSLLLQVSGRKQLLLGQFTLLFNPWNREDAVFLKNEAQRMEYLLNQNGLIYLGTADCIQAESWDFGQFEGDVIDLSLRLLSKDKQVEKWSQPVHVARVLGALLHFLKEQRVLPTPQTQATQEGALLNKRRGSVPILRQWLTGRGRPVYDGQAWVLAAVACTVLRCLGIPARVVTTFASAQGTGGRLLIDEYYNEEGLQNGEGQRGRIWKCCEDGTLELTDSNTKYVGNNISTKGVGSDRCEDITQNYKYPEGSLQEKEVLERVEKEKMEREKDNGIRPPSLETASPLYLLLKAPSSLPLRGDAQISVTLVNHSEQEKAVQLAIGVQAVHYNGVLAAKLWRKKLHLTLSANLEKIITIGLFFSNFERNPPENTFLRLTAMATHSESNLSCFAQEDIAICRPHLAIKMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSASpecifications
erythrocyte membrane protein band 4.2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human EPB42 protein. | |
EPB42 | |
MGC116735/MGC116737/PA | |
Mouse | |
50 μL | |
Yes | |
2038 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC099627.1 | |
EPB42 | |
EPB42 (AAH99627.1, 1 a.a. ∼ 619 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |