Få mere at vide
ENO1, Human, Matched Antibody Pair (AP52), Abnova™
Beskrivelse
Indeholder
Capture antibody: mouse monoclonal anti-ENO1, IgG1 Kappa (100μg) , Detection antibody: rabbit purified polyclonal anti-ENO1 (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Tekniske data
Tekniske data
| Antigen | ENO1 |
| Applikationer | ELISA |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ENO1. |
| Gene | ENO1 |
| Gene Alias | ENO1L1/MBP-1/MPB1/NNE/PPH |
| Gensymboler | ENO1 |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only