missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial membrane protein 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant EMP1.
Brand: Abnova H00002012-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MLVLLAGIFVVHIATVIMLFVSTIANVWLVSNTVDASVGLWKNCTNISCSDSLSYASEDALKTVQAFMILSIIFCVIALLVFVFQLFTMEKGNRFFLSGATTLVCWLCILVGVSIYTSHYANRDGTQYHHGYSYILGWICFCFSFIIGVLYLVLRKKSpecifications
epithelial membrane protein 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant EMP1. | |
EMP1 | |
CL-20/EMP-1/TMP | |
Mouse | |
50 μL | |
Yes | |
2012 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC047300 | |
EMP1 | |
EMP1 (AAH47300, 1 a.a. ∼ 157 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |