missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dynorphin B Rabbit anti-Guinea Pig, Hamster, Non-human primate, Porcine, Rat, Polyclonal, Invitrogen™
Beskrivelse
Predicted to react with bovine, chimpanzee, dog, gorilla, horse, human and mouse based on sequence homology.
The protein encoded by this gene is a preproprotein that is proteolytically processed to form the secreted opioid peptides beta-neoendorphin, dynorphin, leu-enkephalin, rimorphin, and leumorphin. These peptides are ligands for the kappa-type of opioid receptor. Dynorphin is involved in modulating responses to several psychoactive substances, including cocaine.
Tekniske data
Tekniske data
| Antigen | Dynorphin B |
| Applikationer | Immunohistochemistry (Frozen), Radioimmune Assays (RIA) |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Gene | PDYN |
| Genadgangsnr. | Q60478, P01213, P01214, P06300 |
| Gene Alias | MGC26418, PENKB |
| Gensymboler | PDYN |
| Værtsarter | Rabbit |
| Immunogen | Synthetic peptide, YGGFLRRQFKVVT, corresponding to full-length porcine dynorphin B (1-13), conjugated to thyroglobulin. |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?