Learn More
DLC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528443
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Dog (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DLC1 Polyclonal antibody specifically detects DLC1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DLC1 | |
Polyclonal | |
Unconjugated | |
Dlc1 | |
A730069N07Rik; ARHGAP 7; ARHGAP7; deleted in liver cancer 1; deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC1; Dlc-1; DLC1 Rho GTPase activating protein; DLC1 Rho GTPase activating protein; rho GTPase-activating protein 7; FLJ21120; HP; HP protein; KIAA1723; p122 RhoGAP; p122-RhoGAP; rho GTPase activating protein 7; rho GTPase-activating protein 7; RhoGAP; Rho-GTPase-activating protein 7; rho-type GTPase-activating protein 7; STARD 12; STARD12; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; stAR-related lipid transfer protein 12; START domain-containing protein 12; transfer protein 12; unm_sa1141 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10395 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.84 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96QB1 | |
Dlc1 | |
Synthetic peptide corresponding to a region within amino acids 1 and 43 of Human DLC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |