Få mere at vide
CTCF Mouse, Unlabeled, Clone: 48, BD
Beskrivelse
The family of nucleic acid-binding C2H2 type zinc finger transcription factors is divided into two classes. One class consists of small proteins (Gli1, Krox-20, WT1, Egr-1, and Sp1) with conserved zinc finger clusters of 3 to 5 units, while the other class (ZNF91, ZNF74, ZFP37, CTCF) can contain more than 10 zinc finger clusters. CTCF is a ubiquitously expressed, highly conserved transcription factor that contains 10 C2H2- and 1 C2H-type zinc-finger motifs. CTCF binds to and represses transcription at the promoter-proximal regions of c-myc oncogenes,
while CTCF can bind to and activate transcription at the promoter for amyloid β. Another mechanism of CTCF-mediated repression may include binding to insulator regions between enhancers and promoters resulting in enhancer blocking. In addition, CTCF gene may be a candidate tumor suppressor gene, since it localizes to a narrow cancer-associated chromosome region, and has been shown to have tumor-specific rearrangements in breast cancer patients. Thus, CTCF transcriptional regulation may be important for cell cycle progression, differentiation, apoptosis, and tumorigenesis.
Immunofluorescence, Western Blotting
Tekniske data
Tekniske data
| Antigen | CTCF |
| Applikationer | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 48 |
| Koncentration | 250μg/mL |
| Konjugeret | Unconjugated |
| Formulering | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Immunogen | Human CTCF aa. 184-290 |
| Oprensningsmetode | Affinity Purified |
| Vis mere |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.