missing translation for 'onlineSavingsMsg'
Learn More

CRP2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA531036

Additional Details : Weight : 0.01000kg

Product Code. 13435547

  • 3680.00 DKK / 100µL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (88%), Xenopus laevis (87%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CRP2 Polyclonal antibody specifically detects CRP2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CRP2
Polyclonal
Unconjugated
CSRP2
AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein
Rabbit
Antigen affinity chromatography
RUO
1466
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q16527
CSRP2
Recombinant fragment corresponding to a region within amino acids 10 and 193 of Human CSRP2
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers