Få mere at vide
Cre recombinase Rabbit anti-Virus, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519684
Beskrivelse
PA5-19684 targets Cre recombinase in Western blot applications and shows reactivity with Virus samples. The PA5-19684 immunogen is synthetic peptide conjugated to KLH derived from within residues 250 to the C-terminus of Bacteriophage P1 Cre recombinase. PA5-19684 detects Cre recombinase which has a predicted molecular weight of approximately 38 kDa.
Cre-recombinase is a 38 kDa DNA recombinase derived from the P1 bacteriophage. It is highly specific for a 34 bp DNA sequence (loxP) found in P1 DNA. It catalyzes site-specific recombination between two 34-base-pair LOXP sites. Its role is to maintain the phage genome as a monomeric unit-copy plasmid in the lysogenic state. It is homotetramer when bound to DNA and belongs to the "phage" integrase family.
Tekniske data
| Cre recombinase | |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 250 to the C-terminus of Bacteriophage P1 Cre recombinase. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.8 mg/mL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.