Få mere at vide
CDC25C, Human, IP-WB Antibody Pair (PW1), Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit polyclonal anti-CDC25C (300μL), Antibody pair for WB: mouse polyclonal anti-CDC25C (50μL)
Tekniske data
Tekniske data
| Antigen | CDC25C |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | CDC25C |
| Gene Alias | CDC25 |
| Gensymboler | CDC25C |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only