missing translation for 'onlineSavingsMsg'
Få mere at vide

CD80 Mouse anti-Human, PE, Clone: L307.4, BD

Artikelnummer. 15868858
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15868858 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15868858 Leverandør BD Biosciences Leverandørnr. 557227

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Reacts with B7/BB1, a 60kDa transmembrane glycoprotein which was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4+ and CD8+ T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production.

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CD80
Applikationer Flow Cytometry
Klassifikation Monoclonal
Klon L307.4
Konjugeret PE
Oversigt B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7
Formulering Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Human B7-transfected L cells
Oprensningsmetode Affinity Purified
Mængde 100 Tests
Regulatorisk status RUO
Forskningsdisciplin Immunology
Primær eller sekundær Primary
Målarter Human
Indhold og opbevaring Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.