missing translation for 'onlineSavingsMsg'
Få mere at vide

anti-CD62L/L-Selectin PE-Cy7™, Clone: DREG-56, Invitrogen™

CD62L/L-Selectin Monoclonal antibody A16359 from Invitrogen specifically detects CD62L/L-Selectin in Human samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A16359

Produktkode 15236044

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

CD62L/L-Selectin Monoclonal antibody A16359 from Invitrogen specifically detects CD62L/L-Selectin in Human samples. It is validated for Flow Cytometry.

CD62L (L-selectin) is an adhesion glycoprotein that is constitutively expressed on the cell surface of leukocytes and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similarly to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1).
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1, κ
Clone: DREG-56
Immunogen: Human CD62L.
Formulation: 25 Tests Purified Antibody (10mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Uniprot Accession: P14151
Entrez GeneID: 6402
Gene Symbol: SELL
This product detects CD62L/L-Selectin in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Human CD62L.
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

CD62L/L-Selectin
Monoclonal
PE-Cyanine7
P14151
SELL
Human CD62L.
Primary
Human
Purified
Flow Cytometry
DREG-56
25 Tests Purified Antibody (10mg/ml) in proprietary buffer, with 0.09% sodium azide
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Mouse
25 Tests
6402
Store at 4° C
IgG1 κ
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.