missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Monoclonal Antibody (123C3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135249
Additional Details : Weight : 0.01000kg
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
CD56 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P13591 | |
Ncam1 | |
NCAM. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
123C3 | |
Unconjugated | |
Ncam1 | |
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
Mouse | |
Affinity chromatography | |
RUO | |
4684 | |
4° C | |
Liquid |