missing translation for 'onlineSavingsMsg'
Få mere at vide

CD51 Mouse, Unlabeled, Clone: 21, BD

Artikelnummer. 15855759
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15855759 50 μg
15865759 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15855759 Leverandør BD Biosciences Leverandørnr. 611012

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Integrins are a family of proteins that mediate intercellular adhesion or adherence to extracellular matrix proteins. Their roles are essential for embryonic development, tumor metastasis, organ function, and proper immune cell function. Family members are heterodimers that contain a larger α subunit that is unique to each individual receptor and a smaller β subunit that can be shared by several receptors. Based on β subunit content, integrins are divided into the subfamilies β1, β2, and β3. The β3 subfamily contains the vitronectin receptor (αVβ3) and the platelet protein gpIIb/IIIa. The α chain of the vitronectin receptor (VNRα, CD51, integrin αV) has been reported to consist of a disulfide-linked large extracellular (125kDa) subunit and a smaller (25kDa) membrane-anchored subunit. The large subunit contains multiple sequences with homology to calcium-binding sites in other proteins and an RGD-dependent ligand-binding site, while the small subunit contains a transmembrane domain and a short cytoplasmic domain. VNRα has been reported to be noncovalently associated with the VNR β3-chain (CD61, gpIIIa). The VNR mediates cell adhesion to RGD-containing ligands such as vitronectin, von Willebrand factor, fibrinogen, and thrombospondin.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CD51
Applikationer Western Blot
Klassifikation Monoclonal
Klon 21
Koncentration 250μg/mL
Konjugeret Unconjugated
Oversigt Integrin αV; Vitronectin Receptor α; VNRα
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Human CD51/VNRα aa. 609-722
Oprensningsmetode Affinity Purified
Mængde 50 μg
Regulatorisk status RUO
Forskningsdisciplin Immunology
Primær eller sekundær Primary
Målarter Canine, Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG1 κ
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.