missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Monoclonal Antibody (MEM-55), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119461
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes a siliadase-sensitive epitope of CD45RB, a 180-240 kDa single chain type I membrane glycoprotein, variant of CD45 (CD45RB isoform).
CD45RB Monoclonal antibody specifically detects CD45RB in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD45RB | |
Monoclonal | |
APC | |
Ptprc | |
B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5788 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-55 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08575 | |
Ptprc | |
Human thymocytes and T lymphocytes. | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |