Learn More
CD45 Monoclonal Antibody (YAML501.4), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517691
Additional Details : Weight : 0.01000kg
Description
MA5-17691 targets CD45/PTPRC in FACS and WB applications and shows reactivity with Human samples. The MA5-17691 immunogen is rat monoclonal antibody raised against acute myeloid leukemia cells of human origin.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Specifications
CD45 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P08575 | |
PTPRC | |
Rat monoclonal antibody raised against acute myeloid leukemia cells of human origin. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
YAML501.4 | |
PE | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Rat | |
Protein G | |
RUO | |
5788 | |
4° C, do not freeze | |
Liquid |