Learn More
CD45 Monoclonal Antibody (4A8A4C7A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515478
Additional Details : Weight : 0.01000kg
Description
MA5-15478 targets CD45 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15478 immunogen is purified recombinant fragment of CD45 expressed in E. Coli.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
CD45 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P08575 | |
PTPRC | |
Purified recombinant fragment of CD45 expressed in E. Coli. | |
RUO | |
5788 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4A8A4C7A2 | |
Unconjugated | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |