missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529590
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (87%), Rat (88%), Dog (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD44 Polyclonal antibody specifically detects CD44 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD44 | |
Polyclonal | |
Unconjugated | |
CD44 | |
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25406, 960 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P16070, P26051 | |
CD44 | |
Recombinant fragment corresponding to a region within amino acids 1 and 199 of Human CD44 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |