missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Monoclonal Antibody (MEM-59), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119495
Additional Details : Weight : 0.01000kg
Description
CD43 Monoclonal antibody specifically detects CD43 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD43 | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
P16150 | |
SPN | |
Human T lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
MEM-59 | |
Biotin | |
SPN | |
3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
6693 | |
4° C, do not freeze | |
Liquid |