missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD42a Mouse anti-Human, FITC, Clone: ESS, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112214
Additional Details : Weight : 0.01000kg
Description
Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.Specifications
CD42a | |
Monoclonal | |
FITC | |
P14770 | |
GP9 | |
White blood cells. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
ESS | |
GP9 | |
CD42a, GPIX, GP-IX, glycoprotein 9, platelet glycoprotein IX, Platelet GPIX | |
Mouse | |
Protein A/G | |
RUO | |
2815 | |
4° C, do not freeze | |
Liquid |