Få mere at vide
CD40 Mouse anti-Human, PE-Cy7, Clone: 5C3, BD
Beskrivelse
This 5C3 monoclonal antibody specifically binds to CD40, a 45-48 kDa type I integral membrane glycoprotein. CD40 is expressed on B lymphocytes, but is not expressed on terminally differentiated B cells. CD40 is also expressed by endothelial cells, basal epithelial cells and some epithelial cell carcinomas, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, and CD34+ hematopoietic progenitor cells. This antibody is useful for studying the roles played by CD40 in B-cell growth, proliferation, and differentiation including immunoglobulin isotype switching. Anti-CD40 antibodies have been reported to stimulate B-cell proliferation when costimulated with anti-µ, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4.
Tekniske data
Tekniske data
| Antigen | CD40 |
| Applikationer | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 5C3 |
| Konjugeret | PE-Cyanine7 |
| Oversigt | TNFRSF5; TNF receptor superfamily member 5; CD40L receptor; Bp50; p50 |
| Formulering | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Oprensningsmetode | Affinity Purified |
| Mængde | 50 Tests |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.