Learn More
CD3 Monoclonal Antibody (RIV9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17630
Additional Details : Weight : 1.24740kg
Description
MA1-7630 detects CD3 from human, rat and mouse samples. MA1-7630 has been successfully used in ELISA, flow cytometry, immunofluorescence, immunoprecipitation, and immunohistochemistry (frozen) procedures. The MA1-7630 immunogen is a CD3 protein. Store at 4oC short term. For long term storage, store at -80oC.
CD3 Monoclonal antibody specifically detects CD3 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
P04234, P04235, P07766, P09693, P11942, P20963, P22646, P24161 | |
Cd247, Cd3d, CD3E, CD3g | |
CD3, Human white blood cell fraction | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG3 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
RIV9 | |
Unconjugated | |
CD3g | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Mouse | |
Protein G | |
RUO | |
12500, 12501, 12502, 12503, 915, 916, 917, 919 | |
4°C short term, -80°C long term | |
Liquid |