Learn More
CD3e/CD8 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA516606
3063.48 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes canine CD3 epsilon and CD8 cell surface antigens. Clone CA17.2A12 is a FITC-conjugated mouse IgG1 that recognizes canine CD3, a T cell differentiation antigen expressed on mature T lymphocytes. Clone YCATE 55.9 is an RPE-conjugated Rat IgG1 that recognizes canine CD8 epsilon, which is expressed by cytotoxic T cells. YCATE 55.9 binds to CD8 alpha. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD3e/CD8 Cocktail antibody specifically detects CD3e/CD8 in Canine samples. It is validated for Flow Cytometry.Specifications
CD3e/CD8 | |
Cocktail | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P27597, P33706 | |
CD3D, CD3E, CD3G, CD8A, Cd8b | |
Affinity enriched TCR/CD3 membrane proteins isolated from thymocytes and the T cell line CLGL-9, Canine CD8 alpha chimaeric human IgG1 Fc fusion protein. | |
100 Tests | |
Primary | |
Canine | |
Antibody | |
IgG1 |
Flow Cytometry | |
CA17.2A12/YCATE55.9 | |
FITC/PE | |
CD3E | |
AI504783; BB154331; CD3; CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 delta; CD3 epsilon; CD3 gamma-chain; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; Cd3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3epsilon; CD3-epsilon; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD8; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha-chain; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide (p32); CD8 antig | |
Mixed | |
purified | |
RUO | |
403157, 442981, 479419, 483076, 489383 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |