missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3/CD4 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112264
Additional Details : Weight : 0.01000kg
Description
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1) and PE-conjugated CD4 (clone 7E14, isotype IgG1).
CD3/CD4 Cocktail antibody specifically detects CD3/CD4 in Human samples. It is validated for Flow Cytometry.Specifications
CD3/CD4 | |
Cocktail | |
FITC/PE | |
CD247 | |
Cd247; CD247 antigen; CD247 antigen, zeta subunit; CD247 molecule; CD3 antigen delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 gamma-chain; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; Cd3e; CD3E antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3-epsilon; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); CD3-ZETA; CD3zeta chain; Cd4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 r | |
Mouse | |
Protein A/G | |
RUO | |
915, 916, 917, 919, 920 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M2AB, 7E14, 7E14 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P01730, P04234, P07766, P09693, P20963 | |
CD247, CD3D, CD3E, CD3G, CD4 | |
human thymocytes and human peripheral blood T lymphocytes | |
50 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |