Learn More
ITGB1 Monoclonal Antibody (TS2/16), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA2910
Detalles adicionales : Weight : 0.80000kg
Descripción
MA2910 targets CD29 in FACS applications and shows reactivity with Human samples. The MA2910 immunogen is human Beta1 Integrin, the Beta1 chain of the Beta1 integrin heterodimer (CD29). MA2910 detects CD29 which has a predicted molecular weight of approximately 88 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human samples. It is validated for Flow Cytometry.Especificaciones
ITGB1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P05556 | |
ITGB1 | |
Human Beta1 Integrin, the Beta1 chain of the Beta1 integrin heterodimer (CD29) | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
TS2/16 | |
Unconjugated | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein G | |
RUO | |
3688 | |
-20°C | |
Liquid |